Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Assay Percent Range: >99% w/v
Soluble
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | Beige |
| Purity | >99% |
| For Use With (Application) | Laboratory chemicals |
| Quantity | 100 g |
| Product Type | Lipase |
| Form | Powder |
Product Title
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'
