Learn More
Description
MA1-10610 detects Lipase from human samples. MA1-10610 has been successfully used in ELISA applications. The MA1-10610 immunogen is purified human pancreatic Lipase.
Specifications
Specifications
| Antigen | LIPG |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | LIP10-106.4 |
| Concentration | ~1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LIPG |
| Gene Accession No. | Q9Y5X9 |
| Gene Alias | 3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
