Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPG Monoclonal Antibody (LIP10-150.3)
GREENER_CHOICE

Catalog No. 01671565
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-565 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-565 Supplier Invitrogen™ Supplier No. MA110612
  • $600.00
    $497.30 / Each of 1

    Save $102.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10612 detects Lipase from human samples. MA1-10612 has been successfully used in ELISA applications. The MA1-10612 immunogen is purified human pancreatic Lipase.

LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPG
Applications ELISA
Classification Monoclonal
Clone LIP10-150.3
Concentration ∼1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene LIPG
Gene Accession No. Q9Y5X9
Gene Alias 3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719
Gene Symbols LIPG
Host Species Mouse
Immunogen Purified human pancreatic Lipase.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9388
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.