Learn More
Cayman Chemical Lipid Droplets Fluorescence Assay Kit, 480 tests
SDP
Supplier:  Cayman Chemical 500001480TESTS
Encompass_Preferred
Cayman’s Lipid Droplets Fluorescence Assay Kit can be used to study regulators of lipid droplet biogenesis. The main advantage of this assay is that the green fluorescence of Nile Red is both very sensitive and specific for lipid droplets.{17124} Furthermore, changes in lipid droplet biogenesis in various cell types in response to different manipulations can be both qualified and quantified by fluorescence microscopy, flow cytometry, or fluorescent plate readers. Simultaneous visualization of lipid droplets and associated proteins is also possible when used with antibodies conjugated to compatible fluorophores. Oleic acid, which is commonly used to induce lipid droplet formation in cultured cells, is included as a positive control. This kit provides sufficient reagents to effectively treat/stain 480 individual wells of cells when utilized in a 96-well plate format.
Catalog No. NC0258798
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.