Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lipocalin-1 Antibody Pair [HRP]
SDP

Catalog No. NB010349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010349 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010349 Supplier Novus Biologicals™ Supplier No. NBP27965515PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 31.25 pg/mL
Assay Sensitivity 31.25-2000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Lipocalin-1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias lipocalin 1 (tear prealbumin), lipocalin 1-like 2, lipocalin-1, MGC71975, PMFA, Tear lipocalin, Tear prealbumin, Tlc, TPVon Ebner gland protein, VEG protein, VEGPlipocalin 1 (protein migrating faster than albumin, tear prealbumin)
Gene ID (Entrez) 3933
Gene Symbol LCN1
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.25 mg/mL mouse anti-Lipocalin-1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.15 mg/mL of mouse anti-Lipocalin-1 monoclonal antibody.
  • Standard: Each vial contains 35 ng of recombinant Lipocalin-1.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer
    Storage Requirements Storage is content dependent.
    Target Lipocalin-1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 2000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.