Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipophilin B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987325
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7325 Supplier Bioss Supplier No. BS6546R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.

Specifications

Antigen Lipophilin B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SCGB1D2
Gene Accession No. O95969
Gene Alias LIPB; LIPHB; Lipophilin-B; LPHB; LPNB; prostatein-like lipophilin B; SCGB1D2; Secretoglobin family 1D member 2; secretoglobin, family 1D member 2; secretoglobin, family 1D, member 2
Gene Symbols SCGB1D2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Lipophilin B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10647
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.