Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides from Escherichia coli O127:B8, MFCD00212998, 1 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L45161MG
Encompass
BioXtra, suitable for cell culture, γ-irradiated, Synonym: LPS. Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).
Catalog No. NC0736548
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.