Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lipoteichoic Acid Monoclonal Antibody (55)
GREENER_CHOICE

Catalog No. 01676525 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-525 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-525 Supplier Invitrogen™ Supplier No. MA140134
  • $943.00
    $776.65 / Each of 1

    Save $166.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-40134 detects Lipoteichic Acid from gram positive bacterial samples. MA1-40134 has been successfully used in Western blot, ELISA, flow cytometry, immunofluorescence, and Immunohistochemistry (frozen) applications. Positive control: Culture medium of Gram-positive bacteria. Negative control: Culture medium of eukaryotic cells. The MA1-40134 immunogen is a microbial mixture of Streptococcus sobrinus HG961, HG962, HG970, and HG977.

Lipoteichoic acid (LTA) is a glycerol phosphate surface polymer component of the envelope of Gram-positive bacteria. LTA is anchored via its glycolipids to the membrane and carries a polysaccharide chain extending into the peptidoglycan layer of the cell wall. LTA is released spontaneously into the culture medium during growth of gram-positive bacteria. LTA functions as an immune activator with characteristics very similar to lipopolysaccharide (LPS) from Gram-negative bacteria. LTA binds to CD14 and triggers activation predominantly via Toll-like receptor 2. Although LTA is internalized and traffics to the Golgi, the cellular activation in response to LTA occurs at the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lipoteichoic Acid
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 55
Concentration >200 μg/mL
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.02% sodium azide
Gene Alias LTA
Host Species Mouse
Immunogen Microbial mixture of Streptococcus sobrinus HG961, HG962, HG970, and HG977.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.