Learn More
Kyfora Bio LipoVectorTM 25 MG
Supplier:  Kyfora Bio 1449425
Encompass_Preferred
LipoVectorTM is our liposomal transfection reagent designed for the delivery of RNA DNA and proteins into eukaryotic cells (in vitro and in vivo). LipoVectorTM interacts with nucleic acids and the cell membrane to provide efficient and safe delivery into cells. It can be used for transient and stable gene expression as well as co-transfections and is a powerful tool for intracellular delivery of nucleotides.
Catalog No. 50-255-9804
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.