Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Liproxstatin-1 hydrochloride
GREENER_CHOICE

Catalog No. p-7139250
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Additional Apoptosis-related Compounds

Liproxstatin-1 hydrochloride is a potent ferroptosis inhibitor (IC50 = 22 nM). Protects human renal proximal tubule cells from 1S,3R-RSL3 (Cat.No. 6118) -induced cell death in vitro. Inhibits acute renal failure and prolongs survival in Gpx4 knockdown mice. Also mitigates tissue damage in an in vivo hepatic ischemia/reperfusion model.

Specifications

Synonym N-[(3-Chlorophenyl)methyl]spiro[piperidine-4,2′(1′H)-quinoxalin]-3′-amine hydrochloride
Quantity 5 mg
Target Ferroptosis Inhibitors
Molecular Formula C19H21ClN4.HCl
Formula Weight 377.31
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.