Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liquefied Phenol, USP, 89%, Spectrum™ Chemical
SDP

Catalog No. 18603912 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly-Coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-912 6 x 500 mL Poly-Coated Glass Bottle
18-603-911 500 mL Poly-Coated Glass Bottle
18-603-908 4 L Poly-Coated Glass Bottle
18-603-910 4 x 4 L Poly-Coated Glass Bottle
18-603-913 200 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-912 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PH125500MLGLCS6
Add to Cart
Add to Cart

PH125, 108-95-2, C6H6O

Spectrum™ Chemical Liquefied Phenol, USP is commonly used for permanent treatment of ingrown toe and finger nails, a procedure known as a chemical matrixectomy. All Spectrum Chemical USP products USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 108-95-2 , 7732-18-5
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 89%, 11%
Packaging Poly-Coated Glass Bottle
Quantity 6 x 500 mL
UN Number UN2821
Percent Purity 89%
Grade USP
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.