Learn More
MilliporeSigma™ Supelco™ Liquid chromatography Column Fittings Stopcock, Polycarbonate/Acetal Copolymer
Catalog No. 111011021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Polycarbonate, Acetal Copolymer
Catalog No. Material
11-101-1021 Polycarbonate, Acetal Copolymer
1 options
Catalog No. 11-101-1021 Supplier MilliporeSigma™ Supelco™ Supplier No. S739610EA
Add to Cart
Add to Cart
A polycarbonate/acetal copolymer on-off control with female Luer-Lock inlet and male Luer-Lock outlet ports.
TRUSTED_SUSTAINABILITY
Specifications
Description Luer Lock (A)
Material Polycarbonate, Acetal Copolymer