Learn More
Description
Requires
Light bulb, 7.5w
- Qualitative device causes a 7.5w light bulb to glow when water conducts electricity
- Cord on the side cannot contact water, eliminating shock hazards
- Plastic molded body with recessed electrical connections and electrodes
- Attaches to standard ring stand (up to 0.5 in. dia. [12.7mm]) with thumbscrew clamp (sold separately)
- Comes with illustrated instructions
- Weight: 3.88 oz. (0.11kg)
Operates on 110V, AC only
Specifications
Specifications
| For Use With (Application) | Determine the conductivity of liquids or concentrations |
| Type | Conductivity Indicator |
| Voltage | 110 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

