Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Liquid Convection Apparatus
SDPSCI_ED

Catalog No. S111001 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Borosilicate Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S111001 Borosilicate Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111001 Supplier United Scientific™ Supplier No. UNLCA001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Demonstrate convection in a liquid with this simple tool.

  • The liquid convection apparatus is made from Borosilicate Glass and is bent into a rectangle with a filling funnel.
  • To show the convection of heat in a liquid, simply fill it with a colored liquid and place a heat source at one of the bottom corners.
  • Glass tube is 20 mm outer diameter bent into rectangle approximately 200 x 150 mm.
  • Comes in a custom package to ensure product arrives and can be stored safely.
  • Required but not included are a support stand, clamp, heat source and colored liquid.

Specifications

Grade 8 to 12
Material Borosilicate Glass
Requires Support Stand, Clamp, Heat Source, Colored Liquid
Class Size 1 Lab Group
Product Type Heat Experiment Student Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.