Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liquid Crystal, TK-LQ 3858 Electric effect type, Mesomorphic range:38-58deg C [Nematic Liquid Crystal], TCI America™
SDP

Catalog No. T06991G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T06991G Supplier TCI America Supplier No. T06991G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

MDL Number MFCD00148665

Specifications

Color Yellow
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Liquid Crystal, TK-LQ 3858 Electric effect type, Mesomorphic range:38-58°C [Nematic Liquid Cryst
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.