Learn More
Description
- Antibacterial, PH balanced and dermatologist tested for gentle and effective hand washing.
- Formula made in the USA
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Light Blue |
| Form | Gel |
| Disposable | Reusable |
| Latex Free | Yes |
| Sterility | Non-sterile |
| Volume (English) | 16 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.