Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Liquid Skin™ Waterproof Bandages

Catalog No. 19090599 Shop All Medique Products
Change view
Click to view available options
Format:
Sting-free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-090-599 Sting-free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-599 Supplier Medique Supplier No. 92812
Add to Cart
Add to Cart

Easy-to-apply liquid bandage for use on wounds on hard-to-cover areas.

  • Sting-free
  • Forms breathable, protective layer that promotes healing and prevents infection
  • Reduces exposure to dirt and germs

Specifications

For Use With (Application) Use On Hard-To-Cover Wounded Areas
Product Type Waterproof Liquid Bandage
Format Sting-free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.