Learn More
Apexbio Technology LLC Lisinopril 76547-98-3 1g
SDP
Supplier:  Apexbio Technology LLC B22021000
Encompass_Preferred
Lisinopril (CAS 76547-98-3) is a small molecule inhibitor of angiotensin-converting enzyme (ACE) By blocking ACE activity lisinopril reduces the formation of angiotensin II which diminishes vasoconstriction and aldosterone-mediated sodium and water retention resulting in reduced peripheral vascular resistance This compound is frequently utilized in biomedical research to investigate the pathophysiological mechanisms of hypertension congestive heart failure and cardiovascular disorders as well as to develop and validate preclinical pharmacological models related to ACE inhibition
Catalog No. 50-000-91009
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.