Learn More
Medchemexpress LLC HY-18206A 500mg Medchemexpress, Lisinopril (dihydrate) CAS:83915-83-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY18206A500MG
Encompass_Preferred
Medchemexpress, HY-18206A 500mg Lisinopril (dihydrate) CAS:83915-83-7 Lisinopri dihydrate (MK-521 dihydrate) is angiotensin-converting enzyme inhibitor, used in treatment of hypertension, congestive heart failure, and heart attacks. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2025
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.