Learn More
West Chester™ Cotton Lisle Inspection Gloves with Unhemmed Cuff
Catalog No. 19821575 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
Catalog No.
19-821-575 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-821-575 Supplier West Chester™ Supplier No. 70514
This item is not returnable. View return policy
Constructed in a two piece, reversible pattern using 100% cotton fabric. The longer, 14 in. length is ideal for inspection and electronics assembly.
  • Designed to minimize contamination of products from human hands
  • Can be used as liners under disposable and unsupported liquid proof gloves
  • Reversible for a better protection and comfort
  • Snug fit for excellent dexterity
  • Effective as a glove liner
Specifications
Construction String Knit
Description Regular Weight
Color White
Hand Ambidextrous
Material Cotton
Product Type Inspection Glove
Size Large
Style Reversible
Glove Cuff Color White
Glove Cuff Style Unhemmed
Thumb Style Wing
Length (English) 14 in.
Length (Metric) 35.6 cm