Learn More
PIP™ CleanTeam™ Heavyweight Cotton Lisle Inspection Gloves with Overcast Hem
Catalog No. 19152936 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Men's
Catalog No.
19-152-936 Men's
1 options
This item is not returnable. View return policy
Catalog No. 19-152-936 Supplier PIP™ Supplier No. 97540H
This item is not returnable. View return policy
Constructed in a two piece, reversible pattern using 100% cotton fabric. Designed to minimize contamination of products from human hands, ideal for inspection and electronics assembly.
  • Can be used as liners under disposable and unsupported liquid proof glove
  • Made from component materials which comply with Federal regulations for food contact
  • Overcast hem shed less lint
  • Silicone free
Specifications
Certifications/Compliance 21 CFR, 170-199
Construction String Knit
Coating Material Uncoated
Color White
Disposable Limited-use
Hand Ambidextrous
Latex Free Yes
Material Cotton
Product Type Inspection Gloves
Size Universal
Sterility Non-sterile
Style Reversible
Gender Men's
Glove Cuff Style Overcast Hem
Length (English) 8.7 in.
Length (Metric) 22 cm