Learn More
Microbiologics™ Listeria monocytogenes derived from ATCC 13932, Epower
Shop All Microbiologics Products

Description
- Biosafety Level: 2
- 10 pellets of a single microorganism with a predetermined quantitative assay
- Serotype 4b
- Mean Assay Value: 1.0-9.9E+03 CFU per pellet
Specifications
Specifications
| ATCC® | ATCC 13932 |
| Biosafety Level | 2 |
| Content And Storage | 2°C to 8°C |
| Format | Epower |
| Manufacturing Origin | United States |
| Product Type | QC Microorganism |
| For Use With (Application) | Media Testing |
| Packaging Type | Plastic Canister |
| Quantity | 10 Pellets |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.