Learn More
Mopec™ Liston Bone Cutting Angled Forceps
SDP
Catalog No. 22445730 Shop All Mopec Products
Change view
Click to view available options
Length (English):
5.5 in.
7.5 in.
10 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-445-730 5.5 in.
22-445-751 10 in.
22-445-782 7.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-445-730 Supplier Mopec™ Supplier No. AG052
Add to Cart
Add to Cart
Effective bone cutting instrument for use during autopsies and post-mortem dissection. Sharp tips perform a clean extraction.
  • Complete with smooth grip
Specifications
Autoclavable Not Autoclavable
Color Silver
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Length (English) 5.5 in.