Learn More
Medchemexpress LLC HY-124733A 5mg Medchemexpress, LIT-001 CAS:2245072-21-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124733A5MG
Encompass_Preferred
Medchemexpress, HY-124733A 5mg LIT-001 CAS:2245072-21-1 LIT-001 is the first nonpeptide oxytocin receptor (OT-R) agonist (EC50=55 nM; Ki=226 nM). LIT-001 improves social interaction in a mouse model of autism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9680
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.