Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium 2-ethylhexanoate

Catalog No. AA3933009
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3933009 10 g
AA3933004 2 g
AA3933018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3933009 Supplier Thermo Scientific Chemicals Supplier No. 03933009
Add to Cart

CAS: 15590-62-2 | C8H15LiO2 | 150.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15590-62-2
Molecular Formula C8H15LiO2
Molecular Weight (g/mol) 150.146
MDL Number MFCD00058702
InChI Key MAZKNBXUMANNDL-UHFFFAOYSA-M
Synonym lithium 2-ethylhexanoate, 2-ethylhexanoic acid lithium salt, hexanoic acid, 2-ethyl-, lithium salt 1:1, hexanoic acid, 2-ethyl-, lithium salt, acmc-20akn3, lithium 1+ ion 2-ethylhexanoate, lithium 1+ 2-ethylhexanoate, hexanoic acid,2-ethyl-, lithium salt 1:1
PubChem CID 23672311
IUPAC Name lithium;2-ethylhexanoate
SMILES [Li+].CCCCC(CC)C(=O)[O-]

Specifications

Quantity 10 g
Formula Weight 150.15
Physical Form Lumps
Chemical Name or Material Lithium 2-ethylhexanoate
Hazard Category H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 239-657-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.