Learn More
Sigma Organic Chemistry Lithium 2-hydroxybutano | 1G | MFCD00040991
Supplier: Sigma Organic Chemistry CDS0004571G

Lithium 2-hydroxybutano , 1G
About this Item:
MDL #: MFCD00040991
Molecular Weight: 110.04
UNSPSC Code: 12352200
Chemical Formula: C4H7LiO3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.