Learn More
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS | 653-14-5 |
| Physical Form | Neat |
| Linear Formula | C7H3I2 LiO3 |
| MDL Number | MFCD00002443 |
| Synonym | 2-Hydroxy-3,5-diiodobenzoic acid lithium salt; 3,5-Diiodo-2-hydroxybenzoic acid lithium salt; 3,5-Diiodosalicylic acid lithium salt; LIS |
| Type | Analytical Standard |
| Quantity | 25 g |
| Formula Weight | 395.85 |
| Grade | Analytical Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.