Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium acetate dihydrate, 98%, for analysis

Catalog No. p-4813511
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC447712500 250 g Plastic Bottle
AC447710010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC447712500 Supplier Thermo Scientific Chemicals Supplier No. 447712500
Only null left
Add to Cart

CAS: 6108-17-4 | C2H3LiO2·2H2O | 102.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H3LiO2·2H2O
Molecular Weight (g/mol) 102.02
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym lithium acetate dihydrate, lithiumacetatedihydrate, lithium acetate dihydrate, reagent grade, lioac-2h2o, lioac.2h2o, ksc498a1d, c2h3o2.li.2h2o, lithotab acetate ion dihydrate, acetic acid lithium salt dihydrate
PubChem CID 23666338
IUPAC Name lithium;acetate;dihydrate
SMILES [Li+].CC(=O)[O-].O.O

Specifications

Melting Point 56.0°C to 58.0°C
Quantity 250 g
Assay Percent Range 98%
Linear Formula CH3CO2Li·2H2O
Merck Index 15,5577
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: very soluble in hot water, 215g/L ethanol
Formula Weight 102.02
Percent Purity 98%
Grade Analytical
Packaging Plastic Bottle
Chemical Name or Material Lithium Acetate Dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.