Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Lithium Battery Absorbent Mat Pad
SDP

Catalog No. 17987838 Shop All New Pig Corporation Products
Change view
Click to view available options
Packaging Quantity:
50 Pk.
8 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
17-987-838 50 Pk.
17-987-839 8 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-838 Supplier New Pig Supplier No. MAT3102
Add to Cart
Add to Cart

Catch drips and leaks from lithium-ion cells using this chemical-resistant mat pad designed to safely absorb lithium-ion battery electrolyte

  • Chemical-Resistant Material:Specifically engineered to safely absorb the volatile and hazardous electrolytes found in lithium-ion batteries without degrading or causing a dangerous reaction.
  • Durable Construction: Made from multiple layers of thermally bonded polypropylene, the pads are tear-resistant and won't rip, fray, or fall apart even when fully saturated, ensuring they hold up during use.
  • Clear Identification: The pads are a distinct pink color, a visual cue that clearly marks them for use with hazardous chemicals and lithium-ion battery spills to prevent accidental misuse.
  • Flame-Resistant: In its unused state, the mat is flame resistant and is designed to melt and self-extinguish when exposed to fire, adding a layer of safety during a thermal event.
  • Convenient Pad Format: Unlike rolls, the pre-cut pad format is ideal for catching drips, soaking up small spills, and handling specific cleanup tasks without the need for cutting, which helps reduce waste.

Specifications

Color Pink
Capacity (English) 3.62 gal.
Material Polypropylene
Type Absorbent Mat
Disposable Single-use
Packaging Quantity 50 Pk.
Weight Capacity Lightweight
Weight (English) 5 lb.
Weight (Metric) 2.3 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.