Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Lithium Battery Absorbent Mat Roll
SDP

Catalog No. 17987836 Shop All New Pig Corporation Products
Change view
Click to view available options
Dimensions (L x W):
600 x 14.75 in.
600 x 29.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-987-836 600 x 14.75 in.
17-987-837 600 x 29.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-836 Supplier New Pig Supplier No. MAT3100
Add to Cart
Add to Cart

Catch drips and leaks from lithium-ion cells using this chemical-resistant mat designed to safely absorb lithium-ion battery electrolyte

  • Specifically Designed for Lithium-Ion Batteries: Formulated to safely absorb the hazardous electrolyte released from damaged lithium-ion batteries without degrading or causing a dangerous reaction.
  • Highly Absorbent and Fast-Acting:Fine-fiber construction and exclusive dimple pattern are designed to wick liquids quickly across the surface, providing rapid absorption for faster and safer cleanups.
  • Durable and Long-Lasting: Made from multiple layers of thermally bonded polypropylene.
  • Flame Resistant:Flame resistant and designed to melt and self-extinguish when exposed to fire, adding a layer of safety during a thermal runaway event.
  • Versatile Roll Format:Allows for easy customization, so you can cut off exactly the amount you need to cover a large area or handle a smaller spill, which helps reduce waste and save money.
  • Easy to Identify: The mat's distinct pink color serves as a visual indicator, clearly marking it for use with lithium-ion battery spills to prevent confusion with other absorbent materials.

Specifications

Color Pink
Capacity (English) 2.17 gal.
Material Polypropylene
Type Absorbent Mat
Disposable Single-use
Weight Capacity Lightweight
Dimensions (L x W) 600 x 14.75 in.
Weight (English) 3 lb.
Weight (Metric) 1.4 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.