Learn More
Sigma Aldrich Lithium beta-hydroxypyruvate hydrate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 3369-79-7 |
| Molecular Formula | C3H3LiO4 -+ xH2O |
| Linear Formula | HOCH2COCOOLi -+ aq |
| MDL Number | MFCD10567214 |
| Quantity | 1 g |
| Synonym | beta-Hydroxypyruvic acid lithium salt hydrate; 3-Hydroxy-2-oxopropanoic acid lithium salt; 3-Hydroxy-2-oxopropionic acid lithium salt |
| Molecular Weight (g/mol) | 109.99 (anhydrous basis) |
| Percent Purity | ≥97.0% (calc. based on dry substance, NT) |
| Biological Activity | Respiratory System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.