Learn More
Lithium bis(trimethylsilyl)amide, 20% (ca 1.06M) soln. in THF/ethylbenzene, packaged in resealable septum cap bottle
Description
Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lithium bis(trimethylsilyl)amide is used in generating enolates for preparing lactone precursors, pyranones, and cyclohexanes. It is used to catalyze the addition of phosphine P-H bonds to carbodiimides leading to phosphaguanidines and in a novel three-step synthesis of disubstituted 1,2,5-thiadiazoles. It is also used in the lithiation of ethyl acetate and in the stereospecific Wadsworth-Emmons synthesis of fluoroalkenes from (E)- and (Z)-fluorovinyl sulfones.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 800 mL |
| Linear Formula | [(CH3)3Si]2NLi |
| UN Number | UN2924 |
| Beilstein | 8136861 |
| Flash Point | -17°C |
| Solubility Information | Reacts with water. |
| Formula Weight | 167.33 |
| Concentration or Composition (by Analyte or Components) | 20% (∼1.06 M) soln. in THF/ethylbenzene |
| Sensitivity | Air and moisture sensitive |
| Density | 0.891 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H304-H314-H335-H336-H351 |
| Hazard Statement | GHS H Statement H225-H314-H351-H335 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 223-725-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.