Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate, ≥99%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15253790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15253790 500 g
ICN15253780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15253790 Supplier MP Biomedicals, Inc Supplier No. 0215253790
Add to Cart
Add to Cart

High-purity ACS reagent grade lithium carbonate for laboratory applications.

  • Grade: ACS Reagent
  • Assay: ≥99%
  • Molecular Formula: CLi2O3
  • Molecular Weight: 73.89 g/mol
  • CAS Number: 554-13-2
  • Storage: Room temperature
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 723°C
Color White
pH 11
Physical Form Powder
Quantity 500 g
Assay Percent Range ≥99%
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3586314
Merck Index 15, 5506
Solubility Information Soluble in water
Formula Weight 73.9
Percent Purity ≥99%
Grade Meets ACS reagent chemical requirements
Manufacturing Origin USA
Loss on Drying max. 1%
Odor Odorless
Packaging Poly Bottle
Sensitivity Hygroscopic
Appearance White Powder
Specific Gravity 2.11
Decomposition Information Carbon Oxides; Lithium Oxides
Density 2.11 g/cm³
Chemical Name or Material Lithium Carbonate
Show More Show Less
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.