CAS RN 554-13-2
Lithium carbonate, 99+%, ACS reagent
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, ACS, 99.0% min
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium Carbonate (Powder/Certified ACS), Fisher Chemical
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, 99.999%, (trace metal basis), extra pure
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, 99%, pure
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium, plasma standard solution, Specpure™ Li 1000μg/mL
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium Carbonate, ≥99%, (ACS Reagent Grade), MP Biomedicals
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium Carbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium Carbonate, MP Biomedicals
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, 99%
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium carbonate, 98+%, for analysis
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium Carbonate ACS AR Powder, Macron Fine Chemicals™
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L Synonym: lithium carbonate,dilithium carbonate,lithonate,lithobid,lithane,eskalith,lithotabs,carbonic acid lithium salt,liskonum,lithizine PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium ICP Standard, 10,000 ppm Li in 5% HNO3, Ricca Chemical
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
Lithium AA Standard, 1000 ppm Li in 3% HNO3, Ricca Chemical
CAS: 554-13-2 Molecular Formula: CLi2O3 Molecular Weight (g/mol): 73.89 MDL Number: MFCD00011084 InChI Key: XGZVUEUWXADBQD-UHFFFAOYSA-L PubChem CID: 11125 ChEBI: CHEBI:6504 IUPAC Name: dilithium(1+) carbonate SMILES: [Li+].[Li+].[O-]C([O-])=O
- 1
- 2