Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate ACS AR Powder, Macron Fine Chemicals™

Catalog No. MK5840125 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
POLYSTORMOR
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-5840-125 125 g POLYSTORMOR
MK-5840-500 500 g POLYSTORMOR
MK-5840-212 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-5840-125 Supplier Avantor Supplier No. 584002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Grade: ACS AR
  • Physical Form: Powder
  • Formula: CLi2O3
  • Formula Weight: 73.89 g/mol
  • CAS Number: 554-13-2
  • Meets ACS Reagent Chemical Requirements
  • For laboratory, research, or manufacturing use

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 723°C
Color White
Physical Form Powder
Quantity 125 g
Linear Formula Li2CO3
Solubility Information Slightly Soluble in water
Formula Weight 73.89
Grade Meets ACS reagent chemical requirements
Odor Odorless
Packaging POLYSTORMOR
Product Line Macron Fine Chemicals
Sensitivity Hygroscopic
Appearance White Powder
Chemical Name or Material Lithium Carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.