Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate, Solstice

Catalog No. 6001962 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
100 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-62 1 kg
60-044-768 6 x 1 kg
60-019-61 100 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-019-62 Supplier Solstice Supplier No. 130101KG
Add to Cart
Add to Cart

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Physical Form Solid
Quantity 1 kg
Assay Percent Range ≥99%
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
Formula Weight 73.89g/mol
Percent Purity ≥99%
Grade purum
Manufacturing Origin USA
Packaging Plastic Bottle
Product Line Solstice
Chemical Name or Material Lithium Carbonate
Show More Show Less
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H319
EINECSNumber 209-062-5
RTECSNumber OJ5800000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.