Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate, MP Biomedicals

Catalog No. ICN212612 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212612 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212612 Supplier MP Biomedicals, Inc Supplier No. 0521261280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity lithium carbonate for research and laboratory use.

  • CAS Number: 554-13-2
  • Molecular Formula: CLi2O3
  • Molecular Weight: 73.888 g/mol
  • Melting Point: 723°C
  • Boiling Point: Decomposes at 1,300°C
  • Density: 2.11 g/cu cm
  • Solubility in water: 1.52 wt% at 0°C
  • Usage Statement: For research or further manufacturing use only, not for direct human use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Melting Point 723°C
Quantity 100 g
Linear Formula Li2CO3
Beilstein 3999191
Solubility Information Soluble in water
Formula Weight 73.89
Odor Odorless
Specific Gravity 2.11
Density 2.11 g/cm³
Chemical Name or Material Lithium Carbonate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.