Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003210 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-210 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-210 Supplier Avantor Supplier No. 236201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

  • Grade: BAKER ANALYZED™ A.C.S. Reagent
  • Formula: CLi2O3
  • Formula Weight: 73.89
  • Meets A.C.S. Reagent Chemical Requirements
  • Assay (Li2CO3): 99.4 %
  • Physical Form: Powder
  • For Laboratory, Research or Manufacturing Use

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium(1+) carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.4%
Linear Formula Li2CO3
Merck Index 15, 5556
Solubility Information Slightly Soluble in water
Formula Weight 73.89
Percent Purity 99.4% (acidimetry)
Grade Meets ACS reagent chemical requirements
Manufacturing Origin USA
Odor Odorless
Packaging Poly Bottle
Product Line J.T. Baker
Specific Gravity 1L = 2.11kg
Density 2.11 g/cm³
Assay Acidimetric
Chemical Name or Material Lithium Carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.