Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Carbonate, puriss p.a, ≥99.0% (T), For Microscopy, Solstice

Catalog No. 60047223 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-223 500 g Poly bottle
60-047-222 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-223 Supplier Solstice Supplier No. 62470500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, reagent (for microscopy),≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Boiling Point 1265°C (1013hPa)
Melting Point 550°C
Color Colorless
pH 9 to 11
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0%
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
InChI Formula 1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
Formula Weight 73.89g/mol
Percent Purity ≥99.0% (T)
Concentration or Composition (by Analyte or Components) Chloride: 20 mg/kg, Nitrate: 5 mg/kg, Sulfate: 50 mg/kg, Ca: 50 mg/kg, Cd: 5 mg/kg, Co: 5 mg/kg, Cr: 5 mg/kg, Cu: 5 mg/kg, Fe: 5 mg/kg, K: 50 mg/kg, Mg: 10 mg/kg, Mn: 5 mg/kg, NH4+: 5 mg/kg, Na: 50 mg/kg, Ni: 5 mg/kg, Pb: 5 mg/kg, Zn: 5 mg/kg
Manufacturing Origin Germany
Identification Passes Test
Odor Odorless
Packaging Poly bottle
Product Line Solstice
Density 3.46g/cm3
Assay Titration
Chemical Name or Material Lithium carbonate
Show More Show Less
Hazard Statement P305 + P351 + P338
RTECSNumber 209-062-5
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.