Learn More
Newcomer Supply Lithium Carbonate, Saturated Aqueous, 1 L, Bluing Agent
SDP
Supplier:  Newcomer Supply 12215B
Encompass
Use: Bluing Reagent. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC2101905