Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lithium Chloride, Molecular Biology Grade, Calbiochem™,

Catalog No. 438002250GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-800-2250GM 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-800-2250GM Supplier MilliporeSigma™ Supplier No. 438002250GM

A direct and noncompetitive inhibitor of GSK-3 (Ki = 2 mM) that activates Wnt-signaling

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Assay Percent Range ≥99%
Formula Weight 42.4
CAS 7447-41-8
Molecular Formula ClLi
Solubility Information Easily soluble in cold water
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
Chemical Name or Material Lithium Chloride
Grade by argentometry

Specifications

Quantity 250 g
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES [Li+].[Cl-]
IUPAC Name lithium(1+) chloride
Molecular Weight (g/mol) 42.39
PubChem CID 433294
ChEBI CHEBI:48607
RTECSNumber OJ5950000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.