Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium cyclopentadienide, 97%

Catalog No. AA3932914
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3932914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3932914 Supplier Thermo Scientific Chemicals Supplier No. 03932914

CAS: 16733-97-4 | C5H5Li | 72.04 g/mol

Lithium cyclopentadienide is a useful reagent in preparing metallocenes and cyclopentadienyl derivatives. It is also used as intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16733-97-4
Molecular Formula C5H5Li
Molecular Weight (g/mol) 72.04
MDL Number MFCD00001439,MFCD00001439
InChI Key CSOHRTAOCDVTQU-UHFFFAOYSA-N
Synonym lithium cyclopentadienide, cyclopentadienyllithium, acmc-1bo1f, lithium cyclopenta-1,3-dien-1-ide, lithium 1+ ion cyclopenta-1,3-dien-1-ide
PubChem CID 14063469
IUPAC Name lithium;cyclopenta-1,3-diene
SMILES [Li+].C1C=CC=[C-]1

Specifications

Quantity 25 g
UN Number UN3180
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 72.04
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Lithium cyclopentadienide
missing translation for 'healthHazard1' H314
missing translation for 'healthHazard2' GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
missing translation for 'healthHazard3' P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.