Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium foil, thickness 0.6 mm, size 25 x 300 mm, purity 99.9% | 7439-93-2 | MFCD00134051 | 1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences GF115979131EA
Lithium foil, thickness 0.6 mm, size 25 x 300 mm, purity 99.9% | Purity: 99.9% | Mol Wt: 6.94 | 7439-93-2 | MFCD00134051 | 1EA