Learn More
Sigma Aldrich Fine Chemicals Biosciences Lithium hydroxide monohydr2KG
Supplier: Sigma Aldrich Fine Chemicals Biosciences 40297425G/DEL
Lithium hydroxide monohydrate is a lithium salt. Crystal structure study reveals it has monoclinic structure with space group C2/m. Its vibrational spectra has been studied using IR, Raman and inelastic neutron scattering (INS) spectroscopy. It is reported to transform the surface of titania monoliths to form flower like structures.