Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Hydroxide, Monohydrate, Crystal, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18611320 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-320 125 g Amber Glass Bottle
18-611-321 500 g Amber Glass Bottle
18-611-322 2.5 kg Amber Glass Bottle
18-611-323 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-611-320 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1156125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-66-3

Specifications

Boiling Point 920°C
Melting Point 462°C to 470°C
Color White
CAS Max % 1
Physical Form Solid
Quantity 125 g
Assay Percent Range 98.0%
Molecular Formula LiOH⋅H2O
Linear Formula LiOH·H2O
UN Number UN2680
Solubility Information Soluble in water
IUPAC Name Lithium Hydroxide
Molecular Weight (g/mol) 41.96
Formula Weight 41.96
Percent Purity 98%
Grade ACS
Concentration or Composition (by Analyte or Components) Lithium Hydroxide¹H2O: 98.0 %, Lithium Carbonate: 2.0 %, Iron (Fe): 0.002 %, Chloride (Cl): 0.01 %, Sulfates (SO4): 0.05 %, Insoluble Matter: 0.01 %, Heavy Metals (as Pb): 0.002 %
Manufacturing Origin USA
Packaging Amber Glass Bottle
Sensitivity Hygroscopic
Appearance Crystals
Specific Gravity 1.51
Decomposition Information Spontaneous Polymerisation
Density 1.51 g/cm³
Chemical Name or Material Lithium Hydroxide
Show More Show Less

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.