Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium iodide, anhydrous, 99.95% (metals basis)

Catalog No. AA4066622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4066622 100 g
AA4066614 25 g
AA4066636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4066622 Supplier Thermo Scientific Chemicals Supplier No. 04066622
Add to Cart

CAS: 10377-51-2 | ILi | 133.84 g/mol

Used as an electrolyte in high temperature batteries and artificial pacemakers. Phosphor , which is a complex of Iodine is used in neuron detection , in an electrolyte of the dye-sensitized solar cell. It is an electrolyte additive for lithium-sulfur batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-51-2
Molecular Formula ILi
Molecular Weight (g/mol) 133.84
MDL Number MFCD00011092
InChI Key HSZCZNFXUDYRKD-UHFFFAOYSA-M
Synonym lithium iodide, lithium iodide, anhydrous, unii-s6k2xet783, lithium iodide 6lii, lithium 1+ ion iodide, lithium iodide, ultra dry, lithium iodide 7lii, lithiumiodide, lithium monoiodide
PubChem CID 66321
IUPAC Name lithium;iodide
SMILES [Li+].[I-]

Specifications

Boiling Point 1170°C to 1190°C
Melting Point 449°C
Color White
Physical Form Crystalline, Powder
Quantity 100 g
Linear Formula LiI
Merck Index 14,5535
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 133.84
Percent Purity 99.95%
Odor Odorless
Packaging Ampule
Refractive Index 1.955
Sensitivity Hygroscopic; Light sensitive
Density 4.076 g/mL
Assay (metals basis)
Chemical Name or Material Lithium iodide, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-822-5
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.