Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Methoxide, 0.10 Molar in Toluene, Reagents

Catalog No. 01334821
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-821 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-821 Supplier Reagents Holdings Llc Supplier No. CL199000500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lithium Methoxide, 0.10 Molar in Toluene, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

  • Chemical Name: Lithium Methoxide
  • Concentration: 0.10 Molar
  • Solvent: Toluene
  • Grade: Reagent
  • Physical Form: Liquid
  • Boiling Point: 111°C
  • Color: Colorless
  • Packaging: Glass Bottle

Chemical Identifiers

CAS 108-88-3 , 67-56-1
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym Lithium Methylate, Lithium Methanolate
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Boiling Point 111°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Linear Formula CH3OLi
Flash Point 4°C (39.2°F)
Formula Weight 92.14
Grade Reagent
Concentration or Composition (by Analyte or Components) 0.10 M in Toluene
Packaging Glass Bottle
Appearance Colorless Liquid
Specific Gravity 2
Chemical Name or Material Lithium Methoxide, 0.10 Molar in Toluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.