Learn More
Lithium molybdenum oxide, 99+%
Description
Lithium molybdenum oxide is mainly used as an inhibitor in some types of industrial air conditioning. It is used in comparative property studies of battery systems. It has been used as a petroleum cracking catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 705°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 173.83 |
| Percent Purity | ≥99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.66 g/mL |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 236-977-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.