Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium Nitrate, Crystal, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611330 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-330 25 g Amber Glass Bottle
18-611-331 125 g Amber Glass Bottle
18-611-333 500 g Amber Glass Bottle
18-611-335 2.5 kg Amber Glass Bottle
18-611-332 12 kg Poly Pail
18-611-334 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-611-330 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L112025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
  • Grade: Reagent
  • Molecular Formula: LiNO3
  • Molecular Weight: 68.95
  • Assay (LiNO3): 99.0 %
  • Chloride (Cl): 0.02 %
  • Sulfates (SO4): 0.05 %
  • Water: 1.0 %
  • Deliquescent: Keep tightly closed

Chemical Identifiers

CAS 7790-69-4

Specifications

Melting Point 251°C
CAS Max % 1
Physical Form Solid
Quantity 25 g
Assay Percent Range 99.00%
Molecular Formula LiNO3
UN Number UN2722
Merck Index 15, 5529
Synonym Nitric Acid, Lithium Salt
Solubility Information Soluble in water
InChI Formula InChI=1S/Li.NO3/c;2-1(3)4/q+1;-1
SMILES [Li+].[O-][N+](=O)[O-]
IUPAC Name lithium nitrate
Molecular Weight (g/mol) 68.95
Formula Weight 68.95
Percent Purity 99%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Amber Glass Bottle
Sensitivity Deliquescent
Appearance Crystals, Granular
Decomposition Information Nitrogen Oxides (NOx), Lithium Oxides
Chemical Name or Material Lithium Nitrate
Show More Show Less

Caution: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.