Learn More
Sigma Organic Chemistry LITHIUM PHOSPHORUS SULFUR CHLORIDE (LPSCL) POWDER BATTERY GRADE | 10G
Supplier: Sigma Organic Chemistry 91613710G

LITHIUM PHOSPHORUS SULFUR CHLORIDE (LPSCL) POWDER BATTERY GRADE, 10G
About This Item:
Linear Formula: Li6PS5Cl
UNSPSC Code: 12352200
Storage: ROOM TEMPERATURE
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.